Anti-SCD Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-26393G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Stearoyl-CoA desaturase
- Antigen symbol:SCD
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:6319
- Antigen synonyms:PRO1933|stearoyl-CoA desaturase opposite strand|MSTP008|FADS5|SCDOS|delta(9)-desaturase|predicted protein of HQ0998|delta-9-desaturase|delta-9 desaturase|fatty acid desaturase|SCD1|acyl-CoA desaturase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: SCD (stearoyl-CoA desaturase (delta-9-desaturase))
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human