Anti-PGC-1 alpha Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
The PGC1 alpha Antibody from Novus Biologicals is a rabbit polyclonal antibody to PGC1 alpha. This antibody reacts with human, mouse, rat, hamster, squirrel. The PGC1 alpha Antibody has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: PGC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Squirrel
Specifications
- Catalog No:
- NOVUNBP1-04676SS
- NOVUNBP1-04676
- Antigen symbol:
- PGC1A
- PGC1A
- Antigen name:
- Peroxisome proliferator-activated receptor gamma coactivator 1-alpha
- Peroxisome proliferator-activated receptor gamma coactivator 1-alpha
- Antigen synonyms:
- PGC-1v|PGC1A|Ligand effect modulator 6|PPARGC1|PPARGC1A|PGC-1-alpha|Peroxisome proliferator-activated receptor gamma coactivator 1-alpha|LEM6|PGC1
- PGC-1v|PGC1A|Ligand effect modulator 6|PPARGC1|PPARGC1A|PGC-1-alpha|Peroxisome proliferator-activated receptor gamma coactivator 1-alpha|LEM6|PGC1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 10891
- 10891
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
Frequently Bought Together