Order Entry
Spain
ContactUsLinkComponent
Anti-SREBF1 Mouse Monoclonal Antibody (Biotin) [clone: 2A4]
Anti-SREBF1 Mouse Monoclonal Antibody (Biotin) [clone: 2A4]
Catalog # NOVUNB600-582B
Supplier:  Novus Biologicals
Anti-SREBF1 Mouse Monoclonal Antibody (Biotin) [clone: 2A4]
Catalog # NOVUNB600-582B
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Sterol regulatory element binding transcription factor 1
  • Antigen symbol:
    SREBF1
  • Clonality:
    Monoclonal
  • Clone:
    2A4
  • Conjugation:
    Biotin
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Gene ID:
    6720
  • Antigen synonyms:
    sterol regulatory element-binding protein 1,|SREBP1|SRBP1|SREBF1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: SREBF1 (sterol regulatory element binding transcription factor 1)
Clonality: monoclonal
Clone: 2A4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: