Anti-WNK4 Rabbit Polyclonal Antibody (DyLight® 550)
NOVUNB600-284R
- Antibody type:Primary
- Antigen name:WNK lysine deficient Protein kinase 4
- Antigen symbol:WNK4
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:65266
- Antigen synonyms:PHA2B|PRKWNK4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: WNK4 (WNK lysine deficient protein kinase 4)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat