Anti-FOXP3 Rabbit Polyclonal Antibody (DyLight® 488)
NOVUNB600-245G
- Antibody type:Primary
- Antigen name:Forkhead Box P3
- Antigen symbol:FOXP3
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:50943
- Antigen synonyms:AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: FOXP3 (forkhead box P3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse