Anti-BIRC5 Mouse Monoclonal Antibody [clone: 60.11]
Type: Primary
Antigen: BIRC5 (baculoviral IAP repeat containing 5)
Clonality: monoclonal
Clone: 60.11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Rat
- Catalog No:
- NOVUNB500-205
- NOVUNB500-205SS
- Antigen symbol:
- BIRC5
- BIRC5
- Antigen name:
- Baculoviral IAP Repeat Containing 5
- Baculoviral IAP Repeat Containing 5
- Antigen synonyms:
- Apoptosis Inhibitor 4|SVV|TIAP|IAP4|API4|Baculoviral IAP repeat containing 5|EPR 1|Apoptosis inhibitor survivin|BIRC 5
- Apoptosis Inhibitor 4|SVV|TIAP|IAP4|API4|Baculoviral IAP repeat containing 5|EPR 1|Apoptosis inhibitor survivin|BIRC 5
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 60.11
- 60.11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 332
- 332
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together