Anti-SLC7A11 Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNB300-317V
- Antibody type:Primary
- Antigen name:solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11
- Antigen symbol:Solute carrier family 7, member 11
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:23657
- Antigen synonyms:xCT|CCBR1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: SLC7A11 (solute carrier family 7 (anionic amino acid transporter light chain, xc- system), member 11)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human