Anti-BAI1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
NOVUNB110-81586F
- Antibody type:Primary
- Antigen name:adhesion G protein-coupled receptor B1
- Antigen symbol:ADGRB1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Gene ID:575
- Antigen synonyms:GDAIF|BAI1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: BAI1 (brain-specific angiogenesis inhibitor 1)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse