Order Entry
Spain
ContactUsLinkComponent
Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 350) [clone: FDO66Q]
Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 350) [clone: FDO66Q]
Catalog # NOVUNB110-78644UV
Supplier:  Novus Biologicals
Anti-HSD3B1 Mouse Monoclonal Antibody (DyLight® 350) [clone: FDO66Q]
Catalog # NOVUNB110-78644UV
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1
  • Antigen symbol:
    HSD3B1
  • Clonality:
    Monoclonal
  • Clone:
    FDO66Q
  • Conjugation:
    DyLight® 350
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Gene ID:
    3283
  • Antigen synonyms:
    HSDB3A|3BETAHSD|HSD3B|HSDB3|SDR11E1|I
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

Specifications

About this item

Type: Primary
Antigen: HSD3B1 (hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 1)
Clonality: monoclonal
Clone: FDO66Q
Conjugation: DyLight® 350
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: