Anti-LMO2 Mouse Monoclonal Antibody (DyLight® 488) [clone: 1A9-3B11]
NOVUNB110-78626G
- Antibody type:Primary
- Antigen name:LIM Domain Only 2
- Antigen symbol:LMO2
- Clonality:Monoclonal
- Clone:1A9-3B11
- Conjugation:DyLight® 488
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:4005
- Antigen synonyms:T-cell translocation protein 2 |Cysteine-rich protein TTG-2 |TTG2|RBTNL1|LIM domain only protein 2 |RHOM2|RBTN2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: LMO2 (LIM domain only 2 (rhombotin-like 1))
Clonality: monoclonal
Clone: 1A9-3B11
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human