Anti-EGR2 Rabbit Polyclonal Antibody (DyLight® 405)
NOVUNB110-59723V
- Antibody type:Primary
- Antigen name:early growth response 2
- Antigen symbol:EGR2
- Clonality:Polyclonal
- Conjugation:DyLight® 405
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western blot:Yes
- Gene ID:1959
- Antigen synonyms:KROX20|AT591|CMT4E|CMT1D
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: EGR2 (early growth response 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Rabbit
Isotype: none
Reactivity: