Anti-KIAA1524 Mouse Monoclonal Antibody (Biotin) [clone: 2G10]
NOVUNB110-59722B
- Antibody type:Primary
- Antigen name:Cancerous Inhibitor of Protein Phosphatase 2A
- Antigen symbol:CIP2A
- Clonality:Monoclonal
- Clone:2G10
- Conjugation:Biotin
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:57650
- Antigen synonyms:CIP2A|p90 autoantigen|p90|cancerous inhibitor of PP2A|protein CIP2A
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: KIAA1524 (KIAA1524)
Clonality: monoclonal
Clone: 2G10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: