Anti-Spectrin beta 3 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB110-58783R
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Spectrin beta 3
- Clonality:Polyclonal
- Conjugation:DyLight® 550
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Gene ID:6712
- Antigen synonyms:SCAR14|GTRAP41|SCA5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: Spectrin beta 3
Clonality: polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human