Anti-INHAT-2 Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56619
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:INHAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µL
- Pk:0,2 mL
Specifications
About this item
Type: Primary
Antigen: INHAT-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: