Order Entry
Spain
ContactUsLinkComponent
Anti-BAX Rabbit Polyclonal Antibody
Anti-BAX Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56097
Supplier:  Novus Biologicals
undefined
Anti-BAX Rabbit Polyclonal Antibody
Catalog # NOVUNB100-56097
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCL2-associated X protein
  • Antigen symbol:
    BAX
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Western blot:
    Yes
  • Gene ID:
    581
  • Antigen synonyms:
    BCL2L4
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: BAX (BCL2-associated X protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: