Anti-FLT1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: FLT1 (fms-related tyrosine kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
- Catalog No:
- NOVUNB100-527
- NOVUNB100-527SS
- Antigen symbol:
- FLT1
- FLT1
- Antigen name:
- FMS-related tyrosine kinase 1
- FMS-related tyrosine kinase 1
- Antigen synonyms:
- FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
- FLT-1|tyrosine-protein kinase receptor FLT|vascular permeability factor receptor|vascular endothelial growth factor receptor 1|VEGFR-1|tyrosine-protein kinase FRT|fms-like tyrosine kinase 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 2321
- 2321
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together