Order Entry
Spain
ContactUsLinkComponent
Anti-Annexin A10 Rabbit Polyclonal Antibody
Anti-Annexin A10 Rabbit Polyclonal Antibody
Catalog # BSBTPA2077
Supplier:  Boster Bio
undefined
Anti-Annexin A10 Rabbit Polyclonal Antibody
Catalog # BSBTPA2077
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Annexin A10
  • Antigen symbol:
    ANXA10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    Annexin-10|ANX14|Annexin-14|Annexin A10
  • Amino acid number:
    308 - 324
  • Sequence:
    HYKKALLAICAGDAEDY
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Annexin A10 (308-324aa HYKKALLAICAGDAEDY), different from the related mouse sequence by two amino acids, and from the related rat sequence by one amino acid
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Annexin A10(ANXA10) detection. Tested with WB, IHC-P in Human, Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Annexin A10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat