Order Entry
Spain
ContactUsLinkComponent
Anti-ADAM10 Rabbit Polyclonal Antibody
Anti-ADAM10 Rabbit Polyclonal Antibody
Catalog # BSBTPA1535
Supplier:  Boster Bio
undefined
Anti-ADAM10 Rabbit Polyclonal Antibody
Catalog # BSBTPA1535
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ADAM Metallopeptidase Domain 10
  • Antigen symbol:
    ADAM10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    AD18|CD156c|HsT18717|MADM|kuz|RAK|AD10
  • Amino acid number:
    612 - 627
  • Sequence:
    SVQWSRHFSGRTITLQ
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human ADAM10 (612-627aa SVQWSRHFSGRTITLQ).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Disintegrin and metalloproteinase domain-containing protein 10(ADAM10) detection. Tested with WB in Human.

  • Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ADAM10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human