- Antibody type:Primary
- Antigen name:Matrix Metalloproteinase 9
- Antigen symbol:MMP9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:92kDa type IV collagenase)|MMP-9|GELB|92 kDa gelatinase|MANDP2|matrix metalloproteinase 9 (gelatinase B|92 kDa type IV collagenase|type V collagenase|matrix metalloproteinase-9|92kDa gelatinase|macrophage gelatinase|CLG4B
- Amino acid number:689 to 705
- UniProtKB:P14780
- Sequence:NQVDQVGYVTYDILQCP
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MMP9 (689-705aa NQVDQVGYVTYDILQCP), different from the related mouse sequence by two amino acids, and from the related rat sequence by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Matrix metalloproteinase-9(MMP9) detection. Tested with WB in Human;Rat.
Matrix metallopeptidase 9(MMP-9), also known as 92 kDa type IV collagenase, 92 kDa gelatinase or gelatinase B(GELB), is an enzyme that in humans is encoded by the MMP9 gene. Proteins of the matrix metalloproteinase(MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes. Most MMPs are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades type IV and V collagens. Studies in rhesus monkeys suggest that the enzyme is involved in IL-8-induced mobilization of hematopoietic progenitor cells from bone marrow, and murine studies suggest a role in tumor-associated tissue remodeling.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg Thimerosal, 0.05mg NaN3.
WB: The detection limit for MMP9 is approximately 0.5ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Predicted Species: Species predicted to be fit for the product based on sequence similarities.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: MMP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human