Order Entry
Spain
ContactUsLinkComponent
Anti-HIF-2-alpha Rabbit Polyclonal Antibody
Anti-HIF-2-alpha Rabbit Polyclonal Antibody
Catalog # BSBTPA1129-1
Supplier:  Boster Bio
undefined
Anti-HIF-2-alpha Rabbit Polyclonal Antibody
Catalog # BSBTPA1129-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Hypoxia-inducible Factor 2-alpha
  • Antigen symbol:
    HIF2A
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    bHLHe73|ECYT4|HIF2A|HIF-1alpha-like factor|PAS domain-containing protein 2|hypoxia-inducible factor 2-alpha|endothelial PAS domain-containing protein 1|class E basic helix-loop-helix protein 73|EPAS-1|hypoxia-inducible factor 2 alpha|PASD2|member of PAS protein 25-|HLF|MOP2|basic-helix-loop-helix-PAS protein MOP2|HIF-1-alpha-like factor|HIF2-alpha|HIF-2-alpha
  • Amino acid number:
    202 - 240
  • Sequence:
    YNNCPPHSSLCGYKEPLLSCLIIMCEPIQHPSHMDIPLD
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at N-terminus of rat HIF-2-alpha (202-240aa YNNCPPHSSLCGYKEPLLSCLIIMCEPIQHPSHMDIPLD)
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Endothelial PAS domain-containing protein 1 (EPAS1) detection. Tested with WB, IHC-P in Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: HIF-2-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat