Anti-Connexin 43/GJA1 Rabbit Polyclonal Antibody
Catalog # BSBTPA1026-1
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Gap junction Protein alpha 1 43kDa
- Antigen symbol:GJA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:GJA1|Connexin 43|CX43|CXA1|Gap junction 43 kDa heart protein|CXN-43|Gap junction alpha-1 protein
- Amino acid number:351 - 367
- Sequence:HELQPLAIVDQRPSSRA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Connexin 43 (351-367aa HELQPLAIVDQRPSSRA), identical to the related rat and mouse sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Gap junction alpha-1 protein (GJA1) detection. Tested with WB, IHC-P, IHC-F, ICC in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Connexin 43/GJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat