Anti-Tenascin Mouse Monoclonal Antibody [clone: T20]
Catálogo # BSBTMA1094
Proveedor: Boster Bio
Especificaciones
- Antibody type:Primary
- Antigen name:Tenascin C
- Antigen symbol:TNC
- Clonality:Monoclonal
- Clone:T20
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:HXB|TN|MGC167029
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Human tenascin
- Size:100 µg
- Pk:0,1 mg
Especificaciones
Acerca de este artículo
Mouse IgG monoclonal antibody for Tenascin, tenascin N (TNN) detection. Tested with WB, IHC-P, IHC-F, ICC in Human. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: Tenascin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human