Order Entry
Spain
ContactUsLinkComponent
Anti-AMPK alpha 1 Mouse Monoclonal Antibody [clone: 2B7]
Anti-AMPK alpha 1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # ABCAAB110036-100
Supplier:  Abcam
undefined
Anti-AMPK alpha 1 Mouse Monoclonal Antibody [clone: 2B7]
Catalog # ABCAAB110036-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein kinase, AMP-activated, alpha 1 catalytic subunit
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Monoclonal
  • Clone:
    2B7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Seq:
    1286
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal [2B7] to AMPK alpha 1.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: AMPK alpha 1
Clonality: Monoclonal
Clone: 2B7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Rat;Human;African green monkey