Order Entry
Spain
ContactUsLinkComponent
Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5]
Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-HLA-DR Mouse Monoclonal Antibody [clone: TAL 1B5]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB20181-100
  • ABCAAB20181-1MG
  • Antigen symbol:
  • HLA DR
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • TAL 1B5
  • Reactivity:
  • Human
  • Host:
  • Mouse
  • Isotype:
  • IgG1
  • Western blot:
  • Yes
  • ELISA:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together