Order Entry
Spain
ContactUsLinkComponent
 
Anti-Wnt1 Rabbit Polyclonal Antibody
 :  Abcam
 
undefined
Anti-Wnt1 Rabbit Polyclonal Antibody
 :  Abcam

 

  • Catalog No:
  • ABCAAB15251-1
  • ABCAAB15251-500
  • Antigen symbol:
  • WNT1
  • Antigen name:
  • wingless-type MMTV integration site family, member 1
  • Antibody type:
  • Primary
  • Clonality:
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Reactivity:
  • Human, Mouse
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

Frequently Bought Together