Anti-Dystroglycan Goat Polyclonal Antibody
BIRBORB95847-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Dystroglycan
- Antigen symbol:DAG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Western blot:Yes
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1:128000; Immunochemistry: 5 ?g/ml; Western Blot: 0.2-0.6 ?g/ml
Type: Primary
Antigen: Dystroglycan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: