Anti-AIRE Goat Polyclonal Antibody
BIRBORB87566-50
: Biorbyt
- Antibody type:Primary
- Antigen name:autoimmune regulator
- Antigen symbol:AIRE
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage buffer:Tris-buffered saline, pH 7,3, 0,5% BSA, 0,02% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
ELISA: 1:32000; Immunochemistry: 2.5 ?g/ml; RIA: 0.01-0.03 ?g/ml
Type: Primary
Antigen: AIRE (autoimmune regulator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human