Anti-HSPA2 Rabbit Polyclonal Antibody
Catalog # BIRBORB76327-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Heat shock 70kDa protein 2
- Antigen symbol:HSPA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Form:lyophilized
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
0.1-0.5 ?g/ml
Type: Primary
Antigen: HSPA2 (heat shock 70kDa protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: