Anti-IKZF4 Rabbit Polyclonal Antibody
BIRBORB67096-100
: Biorbyt
- Antibody type:Primary
- Antigen name:IKAROS family zinc finger 4 (Eos)
- Antigen symbol:IKZF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage buffer:1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:N-terminal region of mouse IKZF4
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Western Blot: 1:1000-1:5000
Type: Primary
Antigen: IKZF4 (IKAROS family zinc finger 4 (Eos))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: