Anti-TBX21 Rabbit Polyclonal Antibody
Catalog # BIRBORB137878-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:T-box 21
- Antigen symbol:TBX21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:lyophilized
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminal of human TBX21, different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TBX21 (T-box 21)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat