Anti-CYP19A1 Mouse Polyclonal Antibody (RBITC)
Catalog # BIRBORB131215-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:cytochrome P450, family 19, subfamily A, polypeptide 1
- Antigen symbol:CYP19A1
- Clonality:Polyclonal
- Conjugation:RBITC (Rhodamine B Isothiocyanate)
- Host:Mouse
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: CYP19A1 (cytochrome P450, family 19, subfamily A, polypeptide 1)
Clonality: polyclonal
Clone:
Conjugation: RBITC
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: