Anti-ZEB1 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)
Catalog # BIRBORB130230-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:zinc finger E-box binding homeobox 1
- Antigen symbol:ZEB1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy5.5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:100 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human ZEB1/AREB6.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: ZEB1 (zinc finger E-box binding homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat