Anti-LGR5 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy3®)
Catalog # BIRBORB129256-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:leucine-rich repeat containing G Protein-coupled receptor 5
- Antigen symbol:LGR5
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy3®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:100 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: LGR5 (leucine-rich repeat containing G protein-coupled receptor 5)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: