Anti-USP32 Rabbit Polyclonal Antibody
: Biorbyt
Type: Primary
Antigen: USP32 (ubiquitin specific peptidase 32)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
- Catalog No:
- BIRBORB127313-200
- BIRBORB127313-100
- Antigen symbol:
- USP32
- USP32
- Antigen name:
- Ubiquitin specific peptidase 32
- Ubiquitin specific peptidase 32
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- ImmunoChemistry:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage buffer:
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- PBS (without Mg2+ and Ca2+), pH 7,4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together