Anti-ALOX12 Rabbit Polyclonal Antibody (Cy7®)
BIRBORB123442-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Arachidonate 12-lipoxygenase
- Antigen symbol:ALOX12
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Antigen synonyms:12-LOX
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:73 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic peptide derived from human 12 Lipoxygenase.
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: ALOX12 (arachidonate 12-lipoxygenase)
Clonality: polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: