Anti-SOX2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
BIRBORB110424-100
: Biorbyt
- Antibody type:Primary
- Antigen name:SRY (sex determining region Y)-box 2
- Antigen symbol:SOX2
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Rabbit
- Isotype:IgG
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:31.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: