Anti-Influenza A, H1N1, H2N2, H3N2, H5N1 Nucleoprotein Mouse Monoclonal Antibody
Catalog # BIRBORB109454-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Influenza A, H1N1, H2N2, H3N2, H5N1 Nucleoprotein
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Virus
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Influenza A blend.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Influenza A, H1N1, H2N2, H3N2, H5N1 Nucleoprotein
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Virus