Order Entry
Spain
ContactUsLinkComponent
Anti-Mast Cell Tryptase, alpha and beta isoforms Mouse Monoclonal Antibody
Anti-Mast Cell Tryptase, alpha and beta isoforms Mouse Monoclonal Antibody
Catalog # BIRBORB108104-0.2
Supplier:  Biorbyt
Anti-Mast Cell Tryptase, alpha and beta isoforms Mouse Monoclonal Antibody
Catalog # BIRBORB108104-0.2
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mast Cell Tryptase, alpha and beta isoforms
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/mL
  • Shipping temperature:
    Ice
  • Immunogen:
    Human mast cell tryptase purified from human lung tissue.
  • Purification:
    purified by affinity chromatography
  • Size:
    200 μg
  • Pk:
    0,2 mg

Specifications

About this item

Immunochemistry: 1:10000

Type: Primary
Antigen: Mast Cell Tryptase, alpha and beta isoforms
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: