Anti-RBBP8 Rabbit Polyclonal Antibody
Catalog # BIRBORB41492-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Retinoblastoma binding protein 8
- Antigen symbol:RBBP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Amino acid number:452 to 747
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 452 and 747 of CtIP .
- Purification:purified by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:3000; Immunochemistry: 1:100-1:500
Type: Primary
Antigen: RBBP8 (retinoblastoma binding protein 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human