Anti-ADAP1 Rabbit Polyclonal Antibody
100 µl
Catalog # BIRBORB41264-100
Marketsource
Supplier: Biorbyt
Jump to:
Product Details & Documents
Western Blot: 1:500-1:3000
Type: Primary
Antigen: ADAP1 (ArfGAP with dual PH domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Tamaño:100 µl
- Western blot:Yes
- Clonalidad:Polyclonal
- Purification:purified by antigen affinity chromatography
- Concentración:0,44 mg/ml
- Tipo de anticuerpo:Primario
- Inmunógeno:Synthetic peptide contain a sequence corresponding to a region within amino acids 312 and 374 of Human CENTA1.
- Reactividad:Human
- Simbolo antígeno:ADAP1
- Conjugado:Unconjugated
- Huésped:Rabbit
- Shipping temperature:Ice
- Cat. No.:BIRBORB41264-100
- Nombre del antígeno:ArfGAP with dual PH Domains 1
- Temperatura de almacenamiento:Cool
- Número de aminoácidos:312 to 374
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



