Anti-thymic dendritic cell derived factor 1 precursor Rabbit Polyclonal Antibody
100 µg
Catalog # BIRBORB41067-100
Marketsource
Supplier: Biorbyt
Anti-thymic dendritic cell derived factor 1 precursor Rabbit Polyclonal Antibody
Catalog # BIRBORB41067-100
Supplier: Biorbyt
Marketsource
Jump to:
Product Details & Documents
Western Blot: 1:500-1:3000
Type: Primary
Antigen: thymic dendritic cell derived factor 1 precursor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Purification:purified by antigen affinity chromatography
- Antibody type:Primary
- Amino acid number:16 to 281
- Storage temperature:Cool
- Host:Rabbit
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Antigen name:thymic dendritic cell derived factor 1 precursor
- Cat. No.:BIRBORB41067-100
- Reactivity:Human
- Immunogen:Recombinant protein fragment contain a sequence corresponding to a region within amino acids 16 and 281 of Human TMEM59.
- Conjugation:Unconjugated
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7).
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



