Rat IgG Isotype Control (Unconjugated)
Catalog # BIRBORB27742-1
Supplier: Biorbyt
Specifications
- Antibody type:Control
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg
Specifications
About this item
Type: Control
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: