Anti-Cyclic AMP Sheep Polyclonal Antibody
Catalog # BIRBORB26747-0.2
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Cyclic Adenosine Monophosphate
- Antigen symbol:cAMP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Succinylated cAMP conjugated to bovine serum albumin.
- Size:200 μL
- Pk:0,2 mL
Specifications
About this item
ELISA: 1:1000
Type: Primary
Antigen: Cyclic AMP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: