Anti-CD28 Mouse Monoclonal Antibody
BIRBORB26503-100
: Biorbyt
- Antibody type:Primary
- Antigen name:CD28 molecule
- Antigen symbol:CD28
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:PBS containing 2 mg/mL BSA.
- Storage temperature:Cool
- Concentration:25 ug/ml
- Shipping temperature:Ice
- Immunogen:Transfected murine cell line.
- Purification:purified by anion exchange chromatography
- Size:100 test
- Pk:100 Tests
Flow Cytometry: 20 ?l
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human