Anti-DEFB103A Mouse monoclonal antibody
Catalog # BIRBORB26481-10
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:defensin, beta 103A
- Antigen symbol:DEFB103A
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:lyophilized
- Amino acid number:6 to 22
- Storage buffer:50mM Tris, pH 7,4
- Storage temperature:Cool
- Concentration:1 mg/ml (prior to lyophilization)
- Shipping temperature:Ice
- Immunogen:Human synthetic beta-Defensin-3 peptide (LQKYYCRVRGGRCAVLS).
- Purification:purified by Protein G chromatography
- Size:10 µg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: DEFB103A (defensin, beta 103A)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human