Anti-Thyroxine (T4) Mouse Monoclonal Antibody
BIRBORB24332-1
: Biorbyt
- Antibody type:Primary
- Antigen name:Thyroxine (T4)
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:2,85 mg/ml (OD280)
- Shipping temperature:Ice
- Immunogen:Thyroxine-BSA
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Thyroxine (T4)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: