Order Entry
Spain
ContactUsLinkComponent
Anti-HIV-2 gp36 Mouse Monoclonal Antibody
Anti-HIV-2 gp36 Mouse Monoclonal Antibody
Catalog # BIRBORB24161-100
Supplier:  Biorbyt
Anti-HIV-2 gp36 Mouse Monoclonal Antibody
Catalog # BIRBORB24161-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HIV-2 gp36
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Amino acid number:
    390 to 702
  • Storage buffer:
    PBS, pH 7,4
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant gp36, amino acids 390-702
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

  • Western Blot: 1:1000; ELISA: 1:500

Type: Primary
Antigen: HIV-2 gp36
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: