Anti-K. pneumoniae 204 Mouse Monoclonal Antibody
BIRBORB23451-1
: Biorbyt
- Antibody type:Primary
- Antigen name:K. pneumoniae 204
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:10,4 mg/ml (OD280, E0,1%=1,4)
- Shipping temperature:Ice
- Immunogen:Lyophilized Klebsiella pneumoniae 204
- Purification:purified by Protein G chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: K. pneumoniae 204
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: