Order Entry
Spain
ContactUsLinkComponent
Anti-Herpes simplex Type-1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anti-Herpes simplex Type-1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BIRBORB23103-1
 :  Biorbyt
Anti-Herpes simplex Type-1 Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
  BIRBORB23103-1
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Herpes simplex Type-1
  • Clonality:
    Polyclonal
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Goat
  • Storage buffer:
    PBS, pH 7,2 containing 10 mg/mL BSA.
  • Storage temperature:
    Cool
  • Concentration:
    1-2 mg/ml (OD280, E(0,1%)=1,4)
  • Shipping temperature:
    Ice
  • Immunogen:
    HSV Type 1, Strain F (Human) Infected Cell Lysate
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Primary
Antigen: Herpes simplex Type-1
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: