Anti-E. coli (all antigens) Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # BIRBORB23087-1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:E. coli (all antigens)
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Storage buffer:PBS containing 10 mg/mL BSA.
- Storage temperature:Cool
- Concentration:1-2 mg/ml (OD280, E(0,1%)=1,4)
- Shipping temperature:Ice
- Immunogen:Mixture of E. coli serotypes.
- Size:1 ml
- Pk:1 mL
Specifications
About this item
Type: Primary
Antigen: E. coli (all antigens)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: none
Reactivity: